Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042748.1 | XP_017898237.1 | 5 - 6 | 806 | 69 | |
GenBank mRNA | JT668557.1 | - | 3 - 4 | 289 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |