Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102190109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709617.3 | XP_005709674.3 | 1 | 616 | 76 | |
GenBank mRNA | JO602454.2 | - | 1 | 811 | 76 | |
JO623310.2 | - | 1 | 1162 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |