Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050942.1 | XP_017906431.1 | 2 - 3 | 445 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |