Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040776.1 | XP_017896265.1 | 2 - 3 | 1792 | 126 | |
GenBank mRNA | JO392910.2 | - | 3 - 4 | 799 | 126 | |
JO427808.1 | - | 3 - 4 | 871 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |