Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698421.3 | XP_005698478.3 | 1 - 2 | 1138 | 68 | |
GenBank mRNA | JT729324.1 | - | 1 - 2 | 47 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |