Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042167.1 | XP_017897656.1 | 10 - 11 | 1710 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |