Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060225.1 | XP_017915714.1 | 5 - 6 | 1019 | 93 | |
GenBank mRNA | JO365657.2 | - | 3 - 4 | 1255 | 93 | |
JO500843.2 | - | 879 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |