Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062384.1 | XP_017917873.1 | 4 - 5 | 1558 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |