Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGF29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040164.1 | XP_017895653.1 | 8 - 9 | 805 | 93 | |
XM_018040165.1 | XP_017895654.1 | 9 - 10 | 845 | 93 | ||
GenBank mRNA | JO628882.2 | - | 2 - 3 | 332 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |