Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102183400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044110.1 | XP_017899599.1 | 9 - 10 | 892 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |