Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675164.3 | XP_005675221.1 | 2 - 3 | 309 | 120 | |
XM_018046920.1 | XP_017902409.1 | 2 - 3 | 445 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |