Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681606.3 | XP_005681663.3 | 4 - 5 | 1134 | 87 | |
XM_018049451.1 | XP_017904940.1 | 4 - 5 | 1209 | 87 | ||
GenBank mRNA | JT663224.1 | - | 4 - 5 | 1827 | 87 | |
KA332786.1 | - | 1 - 2 | 1145 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |