Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102176354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692202.3 | XP_005692259.1 | 4 - 5 | 353 | 91 | |
GenBank mRNA | JO420428.2 | - | 3 - 4 | 1510 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |