Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695134.3 | XP_005695191.1 | 2 - 3 | 342 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |