Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697525.3 | XP_005697582.1 | 4 - 5 | 502 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |