Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699311.3 | XP_005699368.1 | 18 - 19 | 3980 | 86 | |
XM_018042447.1 | XP_017897936.1 | 18 - 19 | 3967 | 86 | ||
GenBank mRNA | JI861778.1 | - | 16 - 17 | 3364 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |