Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043633.1 | XP_017899122.1 | 4 - 5 | 920 | 81 | |
GenBank mRNA | JO343269.2 | - | 2 - 3 | 240 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |