Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047040.1 | XP_017902529.1 | 5 - 6 | 6949 | 72 | |
XM_018047041.1 | XP_017902530.1 | 5 - 6 | 6937 | 72 | ||
XM_018047042.1 | XP_017902531.1 | 6 - 7 | 6098 | 72 | ||
GenBank mRNA | JT665620.1 | - | 4 - 5 | 388 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |