Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102169288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051811.1 | XP_017907300.1 | 2 - 3 | 289 | 65 | |
GenBank mRNA | JO598774.2 | - | 6 - 7 | 1004 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |