Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102169288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051811.1 | XP_017907300.1 | 3 - 4 | 489 | 86 | |
GenBank mRNA | JO598774.2 | - | 5 - 6 | 804 | 86 | |
KA311590.1 | - | 1 - 2 | 151 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |