Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965991.2 | XP_013821445.1 | 2 - 3 | 635 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |