Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061669.1 | XP_017917158.1 | 5 - 6 | 938 | 107 | |
XM_018061670.1 | XP_017917159.1 | 5 - 6 | 850 | 107 | ||
XM_018061671.1 | XP_017917160.1 | 5 - 6 | 772 | 107 | ||
XM_018061672.1 | XP_017917161.1 | 4 - 5 | 862 | 107 | ||
XM_018061673.1 | XP_017917162.1 | 4 - 5 | 778 | 107 | ||
XM_018061674.1 | XP_017917163.1 | 4 - 5 | 627 | 107 | ||
GenBank mRNA | KA341371.1 | - | 5 - 6 | 909 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |