Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674826.3 | XP_005674883.1 | 1 | 1116 | 65 | |
GenBank mRNA | JO341070.2 | - | 1 | 1132 | 65 | |
JR112624.1 | - | 1 | 1132 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |