Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676642.3 | XP_005676699.1 | 4 - 5 | 1317 | 97 | |
XM_005676643.3 | XP_005676700.1 | 4 - 5 | 1286 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |