Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AADACL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690882.3 | XP_005690939.2 | 3 - 4 | 428 | 78 | |
XM_013970413.2 | XP_013825867.2 | 2 - 3 | 524 | 78 | ||
XM_013970414.2 | XP_013825868.2 | 2 - 3 | 524 | 78 | ||
GenBank mRNA | KA313053.1 | - | 2 - 3 | 1910 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |