Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053358.1 | XP_017908847.1 | 3 - 4 | 553 | 68 | |
GenBank mRNA | JO632331.2 | - | 3 - 4 | 621 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |