Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013970630.2 | XP_013826084.1 | 5 - 6 | 736 | 91 | |
XM_013970631.2 | XP_013826085.1 | 5 - 6 | 808 | 91 | ||
XM_013970632.2 | XP_013826086.1 | 5 - 6 | 674 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |