Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973884.2 | XP_013829338.1 | 2 - 3 | 530 | 100 | |
GenBank mRNA | JO644987.2 | - | 4 - 5 | 779 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |