Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973884.2 | XP_013829338.1 | 4 - 5 | 750 | 93 | |
XM_018039140.1 | XP_017894629.1 | 3 - 4 | 611 | 93 | ||
GenBank mRNA | JI861655.1 | - | 3 - 4 | 529 | 93 | |
JO644987.2 | - | 2 - 3 | 559 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |