Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043461.1 | XP_017898950.1 | 3 - 4 | 588 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |