Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013976420.2 | XP_013831874.1 | 9 - 10 | 3764 | 104 | |
XM_018043749.1 | XP_017899238.1 | 8 - 9 | 3854 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |