Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677630.3 | XP_005677687.1 | 2 - 3 | 1073 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |