Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049681.1 | XP_017905170.1 | 8 - 9 | 1485 | 117 | |
GenBank mRNA | JO380617.2 | - | 7 - 8 | 994 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |