Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684661.3 | XP_005684718.2 | 2 - 3 | 341 | 78 | |
GenBank mRNA | KA344607.1 | - | 1 - 2 | 443 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |