Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064747.1 | XP_017920236.1 | 3 - 4 | 360 | 79 | |
GenBank mRNA | JO630623.2 | - | 3 - 4 | 1391 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |