Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694704.3 | XP_005694761.1 | 2 - 3 | 515 | 89 | |
XM_005694705.2 | XP_005694762.1 | 2 - 3 | 485 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |