Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962526.2 | XP_013817980.2 | 4 - 5 | 629 | 66 | |
XM_018045926.1 | XP_017901415.1 | 5 - 6 | 584 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |