Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041773.1 | XP_017897262.1 | 3 - 4 | 776 | 79 | |
XM_018041783.1 | XP_017897272.1 | 3 - 4 | 940 | 79 | ||
XM_018041789.1 | XP_017897278.1 | 3 - 4 | 731 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |