Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013964097.2 | XP_013819551.1 | 4 - 5 | 628 | 71 | |
GenBank mRNA | KA335133.1 | - | 5 - 6 | 870 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |