Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057863.1 | XP_017913352.1 | 2 - 3 | 777 | 96 | |
GenBank mRNA | JT682555.1 | - | 3 - 4 | 254 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |