Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971973.2 | XP_013827427.1 | 2 - 3 | 620 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |