Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049074.1 | XP_017904563.1 | 4 - 5 | 401 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |