Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064890.1 | XP_017920379.1 | 1 - 2 | 133 | 79 | |
XM_018064891.1 | XP_017920380.1 | 1 - 2 | 133 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |