Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064890.1 | XP_017920379.1 | 3 - 4 | 246 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |