Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061020.1 | XP_017916509.1 | 3 - 4 | 1266 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |