Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXHXorf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013976613.2 | XP_013832067.1 | 5 - 6 | 629 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |