Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055389.1 | XP_017910878.1 | 2 - 3 | 252 | 130 | |
GenBank mRNA | JO375060.2 | - | 1 - 2 | 290 | 130 | |
JO530878.2 | - | 1 - 2 | 301 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |