Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694346.3 | XP_005694403.2 | 1 - 2 | 880 | 107 | |
GenBank mRNA | JO596153.2 | - | 1 - 2 | 469 | 107 | |
JO637735.2 | - | 1 - 2 | 956 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |