Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO80B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686329.3 | XP_005686386.2 | 4 - 5 | 632 | 124 | |
GenBank mRNA | KA327107.1 | - | 4 - 5 | 602 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |