Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060227.1 | XP_017915716.1 | 2 | 538 | 104 | |
XM_018060228.1 | XP_017915717.1 | 2 | 535 | 104 | ||
XM_018060229.1 | XP_017915718.1 | 2 | 531 | 104 | ||
GenBank mRNA | JO618211.2 | - | 2 | 148 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |